• USA Home
  • Search Results

  • gliotoxinfromgliocladiumfimbriatum326396799211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gliotoxin from Gliocladium fimbriatum

1 Products
Image of Gliotoxin from <I>Gliocladium fimbriatum</I>
CAS Number:
Empirical Formula (Hill Notation):
 C13H14N2O4S2
Molecular Weight:
 326.39
 Beilstein/REAXYS Number:
 50675
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Mode of Action
Show More