• USA Home
  • Search Results

  • glipizide445542909461911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glipizide

4 Products
Synonym:   1-Cyclohexyl-3-{4-[2-(5-methylpyrazine-2-carboxamido)ethyl]phenylsulfonyl}urea
Image of Glipizide
CAS Number:
Empirical Formula (Hill Notation):
 C21H27N5O4S
Molecular Weight:
 445.54
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
  Application(s)
  Solubility
  Manufacturer designation
Show More