• USA Home
  • Search Results

  • glucagonlikepeptideiamidefragment736human32976310744451911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glucagon-Like Peptide I Amide Fragment 7-36 human

1 Products
Synonym:   Preproglucagon 78-107 Amide
Image of Glucagon-Like Peptide I Amide Fragment 7-36 human
CAS Number:
Empirical Formula (Hill Notation):
 C149H226N40O45
Molecular Weight:
 3297.63
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More