• USA Home
  • Search Results

  • glutaconicacid13010172402311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glutaconic acid

2 Products
Synonym:   2-Pentenedioic acid
Image of Glutaconic acid
CAS Number:
Linear Formula:
 HOOCCH2CH=CHCOOH
Molecular Weight:
 130.10
 Beilstein/REAXYS Number:
 1759560
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More