• USA Home
  • Search Results

  • glutamicacid147135686011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glutamic acid

2 Products
Synonym:   (S)-2-Aminopentanedioic acid, L-Glutamic acid, Glu
Image of Glutamic acid
CAS Number:
Linear Formula:
 HO2CCH2CH2CH(NH2)CO2H
Molecular Weight:
 147.13
 Beilstein/REAXYS Number:
 1723801
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Physical Form
  Special Grade
Show More