• USA Home
  • Search Results

  • glutaricacidmonomethylesterchloride16459150126411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glutaric acid monomethyl ester chloride

1 Products
Synonym:   Methyl 4-(chloroformyl)butyrate, Methyl 5-chloro-5-oxovalerate, Methyl glutaryl chloride
Image of Glutaric acid monomethyl ester chloride
CAS Number:
Linear Formula:
 ClCO(CH2)3COOCH3
Molecular Weight:
 164.59
 Beilstein/REAXYS Number:
 1762383
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More