• USA Home
  • Search Results

  • glycerolformal104109956911611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycerol formal

2 Products
Image of Glycerol formal
CAS Number:
Empirical Formula (Hill Notation):
 C4H8O3
Molecular Weight:
 104.10
 Beilstein/REAXYS Number:
 103206
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Additive
  Chemical Composition
Show More