• USA Home
  • Search Results

  • glycerolpropoxylateblockethoxylate23026908200211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycerol propoxylate-block-ethoxylate

2 Products
Image of Glycerol propoxylate-<I>block</I>-ethoxylate
CAS Number:
Empirical Formula (Hill Notation):
 C8H22O7
Molecular Weight:
 230.26
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Chemical Composition
  Additive
  Molecular weight
  Density
Show More