• USA Home
  • Search Results

  • glyceryltriacetate2182010276111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glyceryl triacetate

2 Products
Synonym:   1,2,3-Triacetoxypropane, 1,2,3-Triacetylglycerol, Glycerol triacetate, Triacetin
Image of Glyceryl triacetate
CAS Number:
Linear Formula:
 (CH3COOCH2)2CHOCOCH3
Molecular Weight:
 218.20
 Beilstein/REAXYS Number:
 1792353
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Density
Show More