• USA Home
  • Search Results

  • glyceryltrinonadecanoate933562653613011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glyceryl trinonadecanoate

2 Products
Synonym:   TG(19:0/19:0/19:0), Trinonadecanoin
Image of Glyceryl trinonadecanoate
CAS Number:
Empirical Formula (Hill Notation):
 C60H116O6
Molecular Weight:
 933.56
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More