• USA Home
  • Search Results

  • glyceryltrioctanoate4706853823811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glyceryl trioctanoate

1 Products
Synonym:   1,2,3-Tricapryloylglycerol, 1,2,3-Trioctanoylglycerol, Glycerol tricaprylate, Glycerol trioctanoate, Glyceryl tricaprylate, Tricaprylin, Trioctanoin
Image of Glyceryl trioctanoate
CAS Number:
Linear Formula:
 [CH3(CH2)6COOCH2]2CHOCO(CH2)6CH3
Molecular Weight:
 470.68
 Beilstein/REAXYS Number:
 1717202
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Biological Source
Show More