• USA Home
  • Search Results

  • glyceryltrioleate8854312232711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glyceryl trioleate

4 Products
Synonym:   (9Z)9-Octadecenoic acid 1,2,3-propanetriyl ester, 1,2,3-Tri(cis-9-octadecenoyl)glycerol, Glycerol trioleate, Glycerol triolein, Oleic acid triglyceride, Oleic triglyceride, TG(18:1(9Z)/18:1(9Z)/18:1(9Z)), Triolein
Image of Glyceryl trioleate
CAS Number:
Linear Formula:
 (C17H33COOCH2)2CHOCOC17H33
Molecular Weight:
 885.43
 Beilstein/REAXYS Number:
 1718692
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Ice Shipping
  Functional group
  Physical Form
  Biological Source
Show More