• USA Home
  • Search Results

  • glycidylmethacrylate1421510691211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycidyl methacrylate

3 Products
Synonym:   2,3-Epoxypropyl methacrylate, GMA, Methacrylic acid 2,3-epoxypropyl ester
Image of Glycidyl methacrylate
CAS Number:
Empirical Formula (Hill Notation):
 C7H10O3
Molecular Weight:
 142.15
 Beilstein/REAXYS Number:
 2506
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Additive
Show More