• USA Home
  • Search Results

  • glycinamidehydrochloride11054166810611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycinamide hydrochloride

2 Products
Image of Glycinamide hydrochloride
CAS Number:
Linear Formula:
 NH2CH2CONH2 · HCl
Molecular Weight:
 110.54
 Beilstein/REAXYS Number:
 3554199
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More