• USA Home
  • Search Results

  • glycine13c215n780521105702211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycine-13C2,15N

1 Products
Synonym:   13C and 15N Labeled glycine, Aminoacetic acid-13C2,15N, Aminoethanoic acid-13C2,15N
Image of Glycine-<SUP>13</SUP>C<SUB>2</SUB>,<SUP>15</SUP>N
CAS Number:
Linear Formula:
 H215N13CH213CO2H
Molecular Weight:
 78.05
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
Show More