• USA Home
  • Search Results

  • glycine75075640611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycine

18 Products
Synonym:   Aminoacetic Acid
Image of Glycine
CAS Number:
Empirical Formula (Hill Notation):
 C2H5NO2
Molecular Weight:
 75.07
 EC Index Number
 200-272-2
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Document Links
  Allergen
  Organoleptic property
  UV-Absorption Condition
  Purity Grade
  Absorption
  Titertest
  Product Line
  Biological Source
  Anal. Standard Form
  Suitability
  Analyte Suitability
  pH Value
  Loss
  UV-Absorption Values
  Solubility
  Physical Form
  Special Grade
Show More

Product #
Description
G8898 Glycine, for electrophoresis, ≥99%,
G7126 Glycine, ReagentPlus®, ≥99% (HPLC),
410225 Glycine, ACS reagent, ≥98.5%,
50046 Glycine, BioUltra, for molecular biology, ≥99.0% (NT),
G8790 Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%,
76524 Glycine, certified reference material, TraceCERT®,
G7403 Glycine, BioXtra, ≥99% (titration),
G5417 Glycine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture,
50058 Glycine, tested according to Ph Eur,
94119 Glycine, analytical standard, for nitrogen determination according to Kjeldahl method,
W328707 Glycine, 99%, FCC,
PHR1111 Glycine, Pharmaceutical Secondary Standard; Certified Reference Material,
1295800 Glycine, United States Pharmacopeia (USP) Reference Standard,
G0450000 Glycine, European Pharmacopoeia (EP) Reference Standard,
W328712 Glycine, 99%, natural (US), FCC, FG,
GX0205 Glycine, GR ACS,
8.16013 Glycine, Glycine for synthesis. CAS 56-40-6, EC Number 200-272-2, chemical formula H₂NCH₂COOH.,
1.03669 Glycine, granulated EMPROVE® EXPERT Ph.Eur.,BP,JP,USP,