• USA Home
  • Search Results

  • glycinemethylesterhydrochloride12555568079511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycine methyl ester hydrochloride

1 Products
Image of Glycine methyl ester hydrochloride
CAS Number:
Linear Formula:
 NH2CH2COOCH3 · HCl
Molecular Weight:
 125.55
 Beilstein/REAXYS Number:
 3593644
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More