• USA Home
  • Search Results

  • glycitin446404024610411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycitin

3 Products
Synonym:   4′,7-Dihydroxy 6-methoxyisoflavone 7-O-glucoside, 4H-1-Benzopyran-4-one, 7-(β-D-glucopyranosyloxy)-3-(4-hydroxyphenyl)-6-methoxy-, Glycitein 7-O-β-glucoside
Image of Glycitin
CAS Number:
Empirical Formula (Hill Notation):
 C22H22O10
Molecular Weight:
 446.40
 MDL number:
 NACRES
 NA.25
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Manufacturer designation
Show More