• USA Home
  • Search Results

  • glycolicacid76057914111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycolic acid

7 Products
Synonym:   Hydroxyacetic acid
Image of Glycolic acid
CAS Number:
Linear Formula:
 HOCH2COOH
Molecular Weight:
 76.05
 Beilstein/REAXYS Number:
 1209322
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39021303
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Vapor pressure
  Transition temperature
  Potency
  Melting Point
  Density
  Bulk Density
  pH Value
  Application(s)
  Solubility
  Product Line
  Physical Form
  Special Grade
Show More