• USA Home
  • Search Results

  • glycyrrhizicacidammoniumsalt839965395604011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycyrrhizic acid ammonium salt

1 Products
Image of Glycyrrhizic acid ammonium salt
CAS Number:
Empirical Formula (Hill Notation):
 C42H62O16 · NH3
Molecular Weight:
 839.96
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More