• USA Home
  • Search Results

  • glycyrrhizicacidammoniumsaltfromglycyrrhizarootlicorice839965395604011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice)

2 Products
Synonym:   3-O-(2-O-β-D-Glucopyranuronosyl-α-D-glucopyranuronosyl)-18β-glycyrrhetinic acid ammonium salt, Glycyrrhizin
Image of Glycyrrhizic acid ammonium salt from <I>glycyrrhiza root (licorice)</I>
CAS Number:
Linear Formula:
 C42H62O16 · NH3
Molecular Weight:
 839.96
 Beilstein/REAXYS Number:
 3897076
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More