• USA Home
  • Search Results

  • glyoxylicacidsolution740429812411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glyoxylic acid solution

1 Products
Synonym:   Formylformic acid, Oxoethanoic acid
Image of Glyoxylic acid solution
CAS Number:
Linear Formula:
 OHCCOOH
Molecular Weight:
 74.04
 Beilstein/REAXYS Number:
 741891
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More