• USA Home
  • Search Results

  • glyphosate16907107183611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Glyphosate

2 Products
Synonym:   N-(Phosphonomethyl)glycine, Glyphosate
Image of Glyphosate
CAS Number:
Linear Formula:
 (HO)2P(O)CH2NHCH2CO2H
Molecular Weight:
 169.07
 Beilstein/REAXYS Number:
 2045054
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More