• USA Home
  • Search Results

  • goctalactone1422010450711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

γ-Octalactone

3 Products
Synonym:   γ-Octanoic lactone, 4-Hydroxy octanoic acid, Octano-1,4-lactone
Image of γ-Octalactone
CAS Number:
Empirical Formula (Hill Notation):
 C8H14O2
Molecular Weight:
 142.20
 Council of Europe no.:
 2274c
 EC Number:
 FEMA Number:
 2796
 Flavis number:
 10.022
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More