• USA Home
  • Search Results

  • goldiiichloridehydrate33979anhydrousbasis2798877811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gold(III) chloride hydrate

3 Products
Synonym:   Auric chloride, Chloroauric acid, Hydrogen tetrachloroaurate(III) hydrate, Tetrachloroauric(III) acid
Image of Gold(III) chloride hydrate
CAS Number:
Linear Formula:
 HAuCl4 · aq
Molecular Weight:
 339.79 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More