• USA Home
  • Search Results

  • goldiiichloridetrihydrate393831696125411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gold(III) chloride trihydrate

2 Products
Synonym:   Hydrogen tetrachloroaurate(III), Tetrachloroauric(III) acid
Image of Gold(III) chloride trihydrate
CAS Number:
Linear Formula:
 HAuCl4 · 3H2O
Molecular Weight:
 393.83
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Package quantity
  Physical Form
  Assay
Show More