• USA Home
  • Search Results

  • goldnanorods1969798765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gold nanorods

17 Products
Synonym:   Au nanorods, Gold nanorod
Image of Gold nanorods
Empirical Formula (Hill Notation):
 Au
Molecular Weight:
 196.97
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Chemical Composition
  Color
  Functional group
  pH Range
  ID clarifier
  Material
  pH Value
  Additive
  Density
  Absorption
  Composite Dimension
  Particle Dimension
  Concentration
Show More

Product #
Description
716812 Gold nanorods, 10 nm diameter, absorption, 780 nm, dispersion in H2O,
716820 Gold nanorods, 10 nm diameter, absorption, 808 nm, dispersion in H2O,
716839 Gold nanorods, 10 nm diameter, absorption, 850 nm, dispersion in H2O,
716898 Gold nanorods, carboxyl terminated, 10 nm diameter, absorption, 808 nm, dispersion in H2O,
716871 Gold nanorods, amine terminated, 10 nm diameter, absorption, 808 nm, dispersion in H2O,
716928 Gold nanorods, palladium coated, 25 nm diameter, 100 μg/mL in H2O, dispersion,
716936 Gold nanorods, platinum coated, 25 nm diameter, 100 μg/mL in H2O, dispersion,
716901 Gold nanorods, methyl terminated, 10 nm diameter, absorption, 808 nm, dispersion in H2O,
771686 Gold nanorods, 25 nm diameter, absorption, 650 nm, dispersion in H2O,
771651 Gold nanorods, 25 nm diameter, absorption, 600 nm, dispersion in H2O,
771643 Gold nanorods, 25 nm diameter, absorption, 550 nm, dispersion in H2O,
776661 Gold nanorods, 10 nm diameter, absorption, 980 nm, dispersion in H2O,
776653 Gold nanorods, 10 nm diameter, absorption, 900 nm, dispersion in H2O,
776688 Gold nanorods, 10 nm diameter, λ: 1064 nm Amax, dispersion in H2O, absorption, 980 nm,
747998 Gold nanorods, 10 nm diameter, silica coated, absorption, 808 nm, dispersion in H2O,
748005 Gold nanorods, 10 nm diameter, silica coated, absorption, 850 nm, dispersion in H2O,
747971 Gold nanorods, 10 nm diameter, silica coated, absorption, 780 nm, dispersion in H2O,