• USA Home
  • Search Results

  • goldnanourchins1969798765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Gold, nano-urchins

6 Products
Synonym:   Gold nanoparticles, Non-functionalized gold nanoparticles
Empirical Formula (Hill Notation):
 Au
Molecular Weight:
 196.97
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  ID clarifier
  pH Range
  Composite Dimension
  Assay
  Miscellaneous Specifications
  Physical Form
  Particle Dimension
  Concentration
  Absorption
Show More