• USA Home
  • Search Results

  • granisetronhydrochloride3488710700799811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Granisetron hydrochloride

4 Products
Synonym:   1-Methyl-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide hydrochloride
Image of Granisetron hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C18H24N4O · HCl
Molecular Weight:
 348.87
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
Show More