• USA Home
  • Search Results

  • graphenenanoplatelets1201778242511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Graphene nanoplatelets

9 Products
Synonym:   GNPs, xGnP® graphene nanoplatelets-grade C-300
CAS Number:
Empirical Formula (Hill Notation):
 C
Molecular Weight:
 12.01
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  ID clarifier
  Chemical Composition
  Physical Form
  Surface Dimension
  Particle Dimension
  Composite Dimension
  Bulk Density
Show More