• USA Home
  • Search Results

  • graphite1201778242511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Graphite

12 Products
Image of Graphite
CAS Number:
Linear Formula:
 C
Molecular Weight:
 12.01 g/mol
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Greener Alternative Principles
  Chemical Composition
  ID clarifier
  Special Grade
  Composite Dimension
  Particle Dimension
  Assay
Show More

Product #
Description
332461 Graphite, flakes,
282863 Graphite, powder, <20 μm, synthetic,
496596 Graphite, powder, <45 μm, ≥99.99% trace metals basis,
496588 Graphite, powder, <150 μm, 99.99% trace metals basis,
496553 Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis,
496537 Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis,
808067 Graphite, flakes, 99% carbon basis, -325 mesh particle size (≥99%), natural,
808083 Graphite, flakes, 99% carbon basis, -325 mesh particle size (50- 70%), natural, wettable,
808113 Graphite, flakes, 99% carbon basis, +50 mesh particle size (≥80%), natural,
808105 Graphite, flakes, 99% carbon basis, +200 mesh particle size (≥70%), natural,
808091 Graphite, flakes, 99% Carbon, −100 mesh (≥80%), natural,
1.04206 Graphite, fine powder extra pure,