• USA Home
  • Search Results

  • greensepaminesfc18micronhplccolumn1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

GreenSep Amine SFC 1.8 Micron HPLC Column

6 Products
 NACRES
 SB.52
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Chromatography Mode
  Application(s)
  Pore size
  Particle Dimension
  Matrix active group
  Manufacturer designation
Show More