• USA Home
  • Search Results

  • greensepdiolsfc5micronhplccolumn1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

GreenSep Diol SFC 5 Micron HPLC Column

11 Products
 NACRES
 SB.52
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Application(s)
  Pore size
  Particle Dimension
  Matrix active group
  Manufacturer designation
Show More

Product #
Description
ES5196 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm,
ES5197 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3.0 mm,
ES5198 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm,
ES5199 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 15 cm × 20 mm,
ES5200 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 15 cm × 30 mm,
ES5201 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm,
ES5202 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3.0 mm,
ES5203 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm,
ES5204 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm,
ES5205 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 25 cm × 20 mm,
ES5206 GreenSep Diol SFC 5 Micron HPLC Column, 5 μm particle size, L × I.D. 25 cm × 30 mm,