• USA Home
  • Search Results

  • gterpinene136239985411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

γ-Terpinene

6 Products
Synonym:   1-Isopropyl-4-methyl-1,4-cyclohexadiene, p-Mentha-1,4-diene, gamma-Terpinene
Image of γ-Terpinene
CAS Number:
Empirical Formula (Hill Notation):
 C10H16
Molecular Weight:
 136.23
 Beilstein/REAXYS Number:
 2038347
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39010915
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More