• USA Home
  • Search Results

  • guaiacol124149005111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Guaiacol

5 Products
Synonym:   2-Methoxyphenol, Catechol monomethyl ether, Pyrocatechol monomethyl ether
Image of Guaiacol
CAS Number:
Linear Formula:
 (CH3O)C6H4OH
Molecular Weight:
 124.14
 Beilstein/REAXYS Number:
 508112
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Manufacturer designation
  Agency/Method
  Assay
  Physical Form
Show More