• USA Home
  • Search Results

  • guaifenesin198229314111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Guaifenesin

3 Products
Synonym:   3-(o-Methoxyphenoxy)-1,2-propanediol, Glycerol guaiacolate, Guaiacol glyceryl ether, Guaifenesin
Image of Guaifenesin
CAS Number:
Empirical Formula (Hill Notation):
 C10H14O4
Molecular Weight:
 198.22
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Special Grade
  Anal. Standard Form
Show More