• USA Home
  • Search Results

  • guanidinehydrochloride95535001111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Guanidine hydrochloride

7 Products
Synonym:   Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride
Image of Guanidine hydrochloride
CAS Number:
Linear Formula:
 NH2C(=NH)NH2 · HCl
Molecular Weight:
 95.53
 Beilstein/REAXYS Number:
 3591990
 EC Number:
 MDL number:
 NACRES:
 NA.31
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Loss
  Titertest
  Product Line
  Color
  Absorption
  Physical Form
  UV-Absorption Values
  Biological Source
  Special Grade
  pH Value
  Solubility
Show More