• USA Home
  • Search Results

  • guanidinethiocyanate1181659384011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Guanidine thiocyanate

6 Products
Synonym:   Guanidinium rhodanide, Guanidinium thiocyanate
Image of Guanidine thiocyanate
CAS Number:
Linear Formula:
 NH2C(=NH)NH2 · HSCN
Molecular Weight:
 118.16
 Beilstein/REAXYS Number:
 3563461
 EC Number:
 MDL number:
 NACRES:
 NA.31
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  Quality clarifier
  Product Line
  Physical Form
  Application(s)
  Melting Point
  Color
  Biological Source
  Special Grade
  Solubility
Show More