• USA Home
  • Search Results

  • guanosine5diphosphatesodiumsalt443204313922611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Guanosine 5′-diphosphate sodium salt

1 Products
Synonym:   GDP
Image of Guanosine 5′-diphosphate sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C10H15N5O11P2
Molecular Weight:
 443.20
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More