• USA Home
  • Search Results

  • guanosine5monophosphatedisodiumsalthydrate40718anhydrousbasis555012911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Guanosine 5′-monophosphate disodium salt hydrate

1 Products
Synonym:   5′-GMP-Na2, 5′-Guanylic acid disodium salt hydrate
Image of Guanosine 5′-monophosphate disodium salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C10H12N5Na2O8P · xH2O
Molecular Weight:
 407.18 (anhydrous basis)
 Beilstein/REAXYS Number:
 3586801
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More