• USA Home
  • Search Results

  • h7291378447787211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

H-7

1 Products
Synonym:   1-(5-Isoquinolinesulfonyl)-2-methylpiperazine, Isoquinoline-5-sulfonic 2-methyl-1-piperazide
Image of H-7
CAS Number:
Empirical Formula (Hill Notation):
 C14H17N3O2S
Molecular Weight:
 291.37
 Beilstein/REAXYS Number:
 5482740
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More

Product #
Description
I6891 H-7,