• USA Home
  • Search Results

  • h7dihydrochloride3642910893017211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

H-7 dihydrochloride

1 Products
Synonym:   1-(5-Isoquinolinylsulfonyl)-2-methylpiperazine dihydrochloride, Isoquinoline-5-sulfonic 2-methyl-1-piperazide dihydrochloride
Image of H-7 dihydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C14H17N3O2S · 2HCl
Molecular Weight:
 364.29
 Beilstein/REAXYS Number:
 5840763
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More