• USA Home
  • Search Results

  • haloperidol375865286811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Haloperidol

5 Products
Synonym:   4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, 4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone, 4-[4-(p-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone
Image of Haloperidol
CAS Number:
Empirical Formula (Hill Notation):
 C21H23ClFNO2
Molecular Weight:
 375.86
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
  Anal. Standard Form
  Physical Form
  Manufacturer designation
  Special Grade
Show More