• USA Home
  • Search Results

  • hatu3802314889310111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

HATU

2 Products
Synonym:   1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate, N-[(Dimethylamino)-1H-1,2,3-triazolo-[4,5-b]pyridin-1-ylmethylene]-N-methylmethanaminium hexafluorophosphate N-oxide
Image of HATU
CAS Number:
Empirical Formula (Hill Notation):
 C10H15F6N6OP
Molecular Weight:
 380.23
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Melting Point
Show More