• USA Home
  • Search Results

  • hederagenin4727046599611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hederagenin

2 Products
Synonym:   3,23-Dihydroxyolean-12-en-28-acid, Astrantiagenin E, Caulosapogenin, Hederagenic acid, Hederagenol
Image of Hederagenin
CAS Number:
Empirical Formula (Hill Notation):
 C30H48O4
Molecular Weight:
 472.70
 MDL number:
 NACRES:
 NA.25
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More