• USA Home
  • Search Results

  • hematoxylinsolutionmayers1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hematoxylin Solution, Mayer’s

5 Products
Image of Hematoxylin Solution, Mayer’s
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  pH Value
Show More