• USA Home
  • Search Results

  • hemin651941600913511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hemin

2 Products
Synonym:   Chloro(protoporphyrinato)iron(III), Chlorohemin, Chloroprotoporphyrin IX iron(III), Ferriprotoporphyrin IX chloride, Hemin(chloride)
Image of Hemin
CAS Number:
Empirical Formula (Hill Notation):
 C34H32ClFeN4O4
Molecular Weight:
 651.94
 Beilstein/REAXYS Number:
 5229914
 EC Number:
 MDL number:
 NACRES:
 NA.61
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Biological Source
  Assay
Show More