• USA Home
  • Search Results

  • hepes23830736545911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

HEPES

11 Products
Synonym:   4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)
Image of HEPES
CAS Number:
Empirical Formula (Hill Notation):
 C8H18N2O4S
Molecular Weight:
 238.30
 Beilstein/REAXYS Number:
 883043
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  pH Value
  Agency/Method
  Suitability
  UV-Absorption Values
  Biological Source
  Loss
  Absorption
  Product Line
  Special Grade
  Application(s)
  Solubility
  Assay
  Physical Form
Show More

Product #
Description
PHG0001 HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture,
H3375 HEPES, ≥99.5% (titration),
H4034 HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture,
H6147 HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration),
H7523 HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration),
54457 HEPES, BioUltra, for molecular biology, ≥99.5% (T),
RDD002 HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%,
PHR1428 HEPES, Pharmaceutical Secondary Standard; Certified Reference Material,
NIST2181 HEPES, NIST® SRM® 2181,
RES6003H-B7 HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture,
54461 HEPES, ≥99.0% (T),