• USA Home
  • Search Results

  • hepesbuffersolution12345736545911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

HEPES buffer solution

1 Products
Synonym:   HEPES solution, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)
Image of HEPES buffer solution
CAS Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
 eCl@ss
 32129211
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  pH Value
  Density
Show More