• USA Home
  • Search Results

  • hepessodiumsalt260297527739311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

HEPES sodium salt

4 Products
Synonym:   4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt
Image of HEPES sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C8H17N2NaO4S
Molecular Weight:
 260.29
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
  Product Line
  Special Grade
  Absorption
Show More